Order Entry
Poland
ContactUsLinkComponent
Anti-CD38 Mouse Monoclonal Antibody [clone: AT1]
Anti-CD38 Mouse Monoclonal Antibody [clone: AT1]
Catalog # ANTIA250691-100
Supplier:  Antibodies.com
New Product
undefined
Anti-CD38 Mouse Monoclonal Antibody [clone: AT1]
Catalog # ANTIA250691-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Acute lymphoblastic leukemia cells antigen CD38
  • Antigen symbol:
    CD38
  • Clonality:
    Monoclonal
  • Clone:
    AT1
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P28907
  • Antigen synonyms:
    ADPRC 1|ADP-ribosyl cyclase 1|CD38 antigen|ADPRC1|cADPr hydrolase 1|ADP ribosyl cyclase/cyclic ADP-ribose hydrolase|ADP ribosyl cyclase|ADP ribosyl cyclase 1|CD 38
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
  • Molecular weight:
    ~45 kDa (Glycoprotein)/35 kDa (protein core)
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Human T cell line CCRF-CEM
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [AT1] antibody to CD38 for Flow Cytometry and IF with samples derived from Human.

  • Validated applications: Flow cytometry, IF
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: AT1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human