Order Entry
Poland
ContactUsLinkComponent
 
Anti-CD6 Rabbit Recombinant Antibody [clone: C6/2884R]
  ANTIA250543-100
New Product
undefined
Anti-CD6 Rabbit Recombinant Antibody [clone: C6/2884R]
  ANTIA250543-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    CD6
  • Antigen symbol:
    CD6
  • Clonality:
    Recombinant
  • Clone:
    C6/2884R
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P30203
  • Antigen synonyms:
    CD6_HUMAN|CD6 antigen Tp120|T-cell differentiation antigen CD6|CD6 molecule|FLJ44171|T12|T cell differentiation antigen CD6|CD_antigen=CD6|OX52
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
  • Molecular weight:
    90 - 130 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human CD6 protein
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Recombinant rabbit monoclonal [C6/2884R] antibody to CD6 for Flow Cytometry, IF and IHC-P with samples derived from Human.

  • Validated applications: Flow cytometry, IF, IHC-P
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: CD6
Clonality: Recombinant
Clone: C6/2884R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human