Anti-Hsp90 alpha Rabbit Polyclonal Antibody
ANTIA12619-100
New Product
- Antibody type:Primary
- Antigen name:EL52
- Antigen symbol:Hsp90 alpha
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P07900
- Antigen synonyms:Heat shock 86 kDa|heat shock 90kD protein, alpha-like 4|epididymis luminal secretory protein 52|heat shock 90kD protein 1, alpha|HS90A_HUMAN|Heat shock 90kDa protein 1 alpha|Heat shock protein HSP 90-alpha|Heat shock 90kD protein 1, alpha like 4|Heat shock protein 90kDa alpha (cytosolic) class A member 1
- Storage buffer:Supplied in Phosphate buffered saline, pH 7,3, with 50% Glycerol and 0,05% Proclin 300
- Molecular weight:90 kDa
- Sequence:AAKKHLEINPDHSIIETLRQKAEADKNDKSVKDLVILLYETALLSSGFSLEDPQTHANRIYRMIKLGLGIDEDDPTADDTSAAVTEEMPPLEGDDDTSRMEEVD
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 629-732 of human Hsp90a (NP_005339.3)
- Tested applications:ICC/IF
- Purification:Affinity purification.
- Pack type:Plastic vial
- Pk:100 µl
Rabbit polyclonal antibody to Hsp90 alpha for WB and ICC/IF with samples derived from human, Mouse, Rat and Monkey.
- Validated applications: WB, ICC/IF
- Recommended dilutions: WB: 1:500 to 1:1000, ICC/IF: 1:20 to 1:5
Type: Primary
Antigen: Hsp90 alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey