Order Entry
Poland
ContactUsLinkComponent
Anti-NSUN5 Rabbit Polyclonal Antibody
Anti-NSUN5 Rabbit Polyclonal Antibody
Catalog # ANTIA11908-100
Supplier:  Antibodies.com
New Product
undefined
Anti-NSUN5 Rabbit Polyclonal Antibody
Catalog # ANTIA11908-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    FLJ10267
  • Antigen symbol:
    NSUN5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q96P11
  • Antigen synonyms:
    NOP2/Sun domain family, member 5A|NSUN5|NSUN5_HUMAN|NOL1|NOL1R|NOL1/NOP2/Sun domain family member 5|MGC986|NOP2/Sun domain family, member 5|NOL1-related protein
  • Storage buffer:
    Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,01% Thiomersal
  • Molecular weight:
    47 kDa
  • Sequence:
    RPGPASQLPRFVRVNTLKTCSDDVVDYFKRQGFSYQGRASSLDDLRALKGKHFLLDPLMPELLVFPAQTDLHEHPLYRAGHLILQDRASCLPAMLLDPPPGSHVIDACAAPGNKTSHLAALLKNQGKIFAFDLDAKRLASMATLLARAGVSCCELAEEDFLAVSPSDPRYHEVHYILLDPSCSGSGMPSRQLEEPGAGTPSPVRLHALAGFQQRALCHALTFPSLQRLVYSTCSLCQEENEDVVRDALQQNPGAFRLAPALPAWPHRGLSTFPGAEHCLRASPETTLSSGFFVAVIERVEVPR
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 127 - 429 of human NSUN5 (NP_060514.1)
  • Tested applications:
    IHC
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody to NSUN5 for WB and IHC with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC
  • Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:200

Type: Primary
Antigen: NSUN5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat