Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (APC (Allophycocyanin)-Cy7®)
Catalog # ANTIA122097-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Acute lymphoblastic leukemia cells antigen CD38
- Antigen symbol:CD38
- Clonality:Monoclonal
- Clone:HIT2
- Conjugation:APC (Allophycocyanin)-Cy7®
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P28907
- Antigen synonyms:ADPRC 1|ADP-ribosyl cyclase 1|CD38 antigen|ADPRC1|cADPr hydrolase 1|ADP ribosyl cyclase/cyclic ADP-ribose hydrolase|ADP ribosyl cyclase|ADP ribosyl cyclase 1|CD 38
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Human thymocytes in foetus
- Purification:This antibody is conjugated with tandem dye APC-Cyanine 7 under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Glass bottle
- Pk:100 T
Specifications
About this item
Mouse monoclonal [HIT2] antibody to CD38 (APC-Cyanine 7) for Flow Cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: HIT2
Conjugation: APC (Allophycocyanin)-Cy7®
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human