Anti-Podoplanin / gp36 Rabbit Recombinant Antibody [clone: PDPN/8875R]
ANTIA316906-100
New Product
- Antibody type:Primary
- Antigen name:Aggrus
- Antigen symbol:Podoplanin / gp36
- Clonality:Recombinant
- Clone:PDPN/8875R
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q86YL7
- Antigen synonyms:GP 38|Glycoprotein 36|gp 36|HT1A 1|gp36|Glycoprotein 36 KD|GP38|GP 40|GP40
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
- Molecular weight:17 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 1-150) of human PDPN protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Recombinant rabbit monoclonal [PDPN/8875R] antibody to Podoplanin / gp36 for ELISA, IF and IHC-P with samples derived from human.
- Validated applications: ELISA, IF, IHC-P
- Recommended dilutions: IF: 1 to 2 µg/ ml, IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: Podoplanin / gp36
Clonality: Recombinant
Clone: PDPN/8875R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human