Order Entry
Poland
ContactUsLinkComponent
Anti-MZF1 Mouse Monoclonal Antibody [clone: PCRP-MZF1-1E8]
Anti-MZF1 Mouse Monoclonal Antibody [clone: PCRP-MZF1-1E8]
Catalog # ANTIA316742-100
Supplier:  Antibodies.com
New Product
Anti-MZF1 Mouse Monoclonal Antibody [clone: PCRP-MZF1-1E8]
Catalog # ANTIA316742-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Myeloid zinc finger 1
  • Antigen symbol:
    MZF1
  • Clonality:
    Monoclonal
  • Clone:
    PCRP-MZF1-1E8
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P28698
  • Antigen synonyms:
    Zfp98|Zinc finger and SCAN domain containing protein 6|MZF1|MZF1_HUMAN|MZF1B|Zinc finger protein 42|MZF 1|Zinc finger and SCAN domain-containing protein 6|MZF-1
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
  • Molecular weight:
    82,05 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Recombinant fragment (around Amino Acids 37-128) of human MZF1 protein.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [PCRP-MZF1-1E8] antibody to MZF1 for flow cytometry, IF and IHC-P with samples derived from human.

  • Validated applications: Flow cytometry, IF, IHC-P
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ ml, IHC: 1 to 2 µg/ ml

Type: Primary
Antigen: MZF1
Clonality: Monoclonal
Clone: PCRP-MZF1-1E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human