Order Entry
Poland
ContactUsLinkComponent
Anti-HSP90 alpha Mouse Monoclonal Antibody [clone: HSP90AA1/7247]
Anti-HSP90 alpha Mouse Monoclonal Antibody [clone: HSP90AA1/7247]
Catalog # ANTIA316499-100
Supplier:  Antibodies.com
New Product
Anti-HSP90 alpha Mouse Monoclonal Antibody [clone: HSP90AA1/7247]
Catalog # ANTIA316499-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    EL52
  • Antigen symbol:
    HSP90 alpha
  • Clonality:
    Monoclonal
  • Clone:
    HSP90AA1/7247
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P07900
  • Antigen synonyms:
    Heat shock 86 kDa|heat shock 90kD protein, alpha-like 4|epididymis luminal secretory protein 52|heat shock 90kD protein 1, alpha|HS90A_HUMAN|Heat shock 90kDa protein 1 alpha|Heat shock protein HSP 90-alpha|Heat shock 90kD protein 1, alpha like 4|Heat shock protein 90kDa alpha (cytosolic) class A member 1
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
  • Molecular weight:
    90 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Recombinant fragment (around Amino Acids 500-700) of human HSP90AA1 protein. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [HSP90AA1/7247] antibody to HSP90 alpha for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ ml

Type: Primary
Antigen: HSP90 alpha
Clonality: Monoclonal
Clone: HSP90AA1/7247
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human