Anti-G-CSF Mouse Monoclonal Antibody [clone: CSF3/4599]
ANTIA316392-100
New Product
- Antibody type:Primary
- Antigen name:C17orf33
- Antigen symbol:G-CSF
- Clonality:Monoclonal
- Clone:CSF3/4599
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P09919
- Antigen synonyms:Colony stimulating factor 3 (granulocyte)|CSF3_HUMAN|Colony stimulating factor 3|Filgrastim|CSF3|CSF3OS|Csfg|CSF beta|CSF 3
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
- Molecular weight:19 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 1-200) of human CSF3 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [CSF3/4599] antibody to G-CSF for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: G-CSF
Clonality: Monoclonal
Clone: CSF3/4599
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human