Anti-GAD65 Rabbit Recombinant Antibody [clone: GAD2/6488R]
Catalog # ANTIA316368-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:65 kDa glutamic acid decarboxylase
- Antigen symbol:GAD65
- Clonality:Recombinant
- Clone:GAD2/6488R
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG kappa
- Reactivity:Human
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# Q05329
- Antigen synonyms:GAD 65|GAD65|GAD-65|Gad2|DCE 2|GAD 2|GAD-2|DCE2|DCE2_HUMAN
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
- Molecular weight:65 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant human GAD2 (GAD65) protein fragment (around Amino Acids 6-99). The exact sequence is proprietary.
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [GAD2/6488R] antibody to GAD65 for ELISA with samples derived from human.
- Validated applications: ELISA
Type: Primary
Antigen: GAD65
Clonality: Recombinant
Clone: GAD2/6488R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human