Anti-CD38 Rabbit Recombinant Antibody [clone: CD38/8335R]
Catalog # ANTIA316079-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Acute lymphoblastic leukemia cells antigen CD38
- Antigen symbol:CD38
- Clonality:Recombinant
- Clone:CD38/8335R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P28907
- Antigen synonyms:ADPRC 1|ADP-ribosyl cyclase 1|CD38 antigen|ADPRC1|cADPr hydrolase 1|ADP ribosyl cyclase/cyclic ADP-ribose hydrolase|ADP ribosyl cyclase|ADP ribosyl cyclase 1|CD 38
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
- Molecular weight:~45 kDa (Glycoprotein)/35 kDa (protein core)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 200-300) of human CD38. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [CD38/8335R] antibody to CD38 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: CD38
Clonality: Recombinant
Clone: CD38/8335R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human