Anti-CD38 Mouse Recombinant Antibody [clone: rCD38/8334]
Catalog # ANTIA316077-100
Supplier: Antibodies.com
New Product
Specifications
- Typ przeciwciała:Primary
- Nazwa antygenu:Acute lymphoblastic leukemia cells antigen CD38
- Symbol antygenu:CD38
- Klonalność:Recombinant
- Klonuj:rCD38/8334
- Koniugacja:Unconjugated
- Gospodarz:Mouse
- ImmunoChemia:Yes
- Izotyp:IgG1 kappa
- Reaktywność:Human
- Formularz:Liquid
- Identyfikator genu:UniprotID# P28907
- Synonimy antygenów:ADPRC 1|ADP-ribosyl cyclase 1|CD38 antigen|ADPRC1|cADPr hydrolase 1|ADP ribosyl cyclase/cyclic ADP-ribose hydrolase|ADP ribosyl cyclase|ADP ribosyl cyclase 1|CD 38
- Bufor do przechowywania:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
- Waga molekularna:~45 kDa (Glycoprotein)/35 kDa (protein core)
- Temperatura przechowywania:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Stężenie:1 mg/ml
- Temperatura transportu:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 200-300) of human CD38. The exact sequence is proprietary.
- Przetestowane aplikacje:IHC-P
- Oczyszczenie:Protein A/G chromatography
- Rodzaj opakowania:Plastic vial
- Ilość:100 µG
Specifications
About this item
Recombinant mouse monoclonal [rCD38/8334] antibody to CD38 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: CD38
Clonality: Recombinant
Clone: rCD38/8334
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human