Order Entry
Poland
ContactUsLinkComponent
Anti-CCL23 Mouse Monoclonal Antibody [clone: CCL23/4034]
Anti-CCL23 Mouse Monoclonal Antibody [clone: CCL23/4034]
  ANTIA315955-100
New Product
Anti-CCL23 Mouse Monoclonal Antibody [clone: CCL23/4034]
  ANTIA315955-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    C C motif chemokine ligand 23
  • Antigen symbol:
    CCL23
  • Clonality:
    Monoclonal
  • Clone:
    CCL23/4034
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2 kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P55773
  • Antigen synonyms:
    C-C motif chemokine 23|CCL23_HUMAN|CCL 23|Chemokine CC motif ligand 23|ck beta 8|CCL23(30-99)|Chemokine (C C motif) ligand 23|CCL23|C6 beta chemokine
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
  • Molecular weight:
    13 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Recombinant fragment (around Amino Acids 1-120) of human CCL23 protein. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [CCL23/4034] antibody to CCL23 for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ ml

Type: Primary
Antigen: CCL23
Clonality: Monoclonal
Clone: CCL23/4034
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2 kappa
Reactivity: Human