Anti-SPARC Mouse Recombinant Antibody [clone: rOSTN/8527]
Catalog # ANTIA315672-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:AA517111
- Antigen symbol:SPARC
- Clonality:Recombinant
- Clone:rOSTN/8527
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P09486
- Antigen synonyms:Cysteine rich protein|Basement membrane protein 40|Basement-membrane protein 40|BM 40|OI17|BM40|BM-40|MGC128090|hm:zeh0062
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide.
- Molecular weight:38 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 1-200) of human SPARC protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant mouse monoclonal [rOSTN/8527] antibody to SPARC for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: SPARC
Clonality: Recombinant
Clone: rOSTN/8527
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human