Anti-HSP90 alpha Mouse Monoclonal Antibody [clone: HSP90AA1/7247]
Catalog # ANTIA315115-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:EL52
- Antigen symbol:HSP90 alpha
- Clonality:Monoclonal
- Clone:HSP90AA1/7247
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P07900
- Antigen synonyms:Heat shock 86 kDa|heat shock 90kD protein, alpha-like 4|epididymis luminal secretory protein 52|heat shock 90kD protein 1, alpha|HS90A_HUMAN|Heat shock 90kDa protein 1 alpha|Heat shock protein HSP 90-alpha|Heat shock 90kD protein 1, alpha like 4|Heat shock protein 90kDa alpha (cytosolic) class A member 1
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide.
- Molecular weight:90 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 500-700) of human HSP90AA1 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [HSP90AA1/7247] antibody to HSP90 alpha for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: HSP90 alpha
Clonality: Monoclonal
Clone: HSP90AA1/7247
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human