Order Entry
Poland
ContactUsLinkComponent
Anti-STAT5A Mouse Monoclonal Antibody [clone: PCRP-STAT5A-1A11]
Anti-STAT5A Mouse Monoclonal Antibody [clone: PCRP-STAT5A-1A11]
Catalog # ANTIA250063-100
Supplier:  Antibodies.com
New Product
undefined
Anti-STAT5A Mouse Monoclonal Antibody [clone: PCRP-STAT5A-1A11]
Catalog # ANTIA250063-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Ma mMary gland factor
  • Antigen symbol:
    STAT5A
  • Clonality:
    Monoclonal
  • Clone:
    PCRP-STAT5A-1A11
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P42229
  • Antigen synonyms:
    STAT5a|Signal transducer and activator of transcription 5A|STAT5|STAT 5A|MGF|STAT 5|STA5A_HUMAN
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    92 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    STAT5A protein domain
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [PCRP-STAT5A-1A11] antibody to STAT5A for flow cytometry with samples derived from human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells

Type: Primary
Antigen: STAT5A
Clonality: Monoclonal
Clone: PCRP-STAT5A-1A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human