Anti-SCXA Mouse Monoclonal Antibody [clone: PCRP-SCXA-2D11]
ANTIA249961-100
New Product
- Antibody type:Primary
- Antigen name:Basic helix loop helix transcription factor scleraxis
- Antigen symbol:SCXA
- Clonality:Monoclonal
- Clone:PCRP-SCXA-2D11
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q7RTU7
- Antigen synonyms:scleraxis homolog A|bHLHa41|Class A basic helix-loop-helix protein 41|Basic helix-loop-helix transcription factor scleraxis|SCX|bHLHa48|Class A basic helix-loop-helix protein 48|SCXB|SCXA
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:22 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human SCXA protein
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [PCRP-SCXA-2D11] antibody to SCXA for flow cytometry and IF with samples derived from human.
- Validated applications: Flow cytometry, IF
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml
Type: Primary
Antigen: SCXA
Clonality: Monoclonal
Clone: PCRP-SCXA-2D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human