Anti-beta 2 Microglobulin Mouse Monoclonal Antibody [clone: SPM617]
Catalog # ANTIA249749-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:B2M
- Antigen symbol:beta 2 Microglobulin
- Clonality:Monoclonal
- Clone:SPM617
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Non-human primates,Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P61769
- Antigen synonyms:CDABP0092|Hdcma22p|Beta chain of mhc class 1 proteins|Beta chain of MHC class I molecules|Beta 2 microglobulin precursor|beta 2 Microglobulin|Beta-2-microglobulin form pI 5.3|Beta 2 microglobin|B2MG_HUMAN
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:12 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human B2M protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [SPM617] antibody to beta 2 Microglobulin for WB, flow cytometry, IF and IHC-P with samples derived from human and non-human primates.
- Validated applications: WB, Flow cytometry, IF, IHC-P
- Recommended dilutions: WB: 1 to 2 µg/ml, Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: beta 2 Microglobulin
Clonality: Monoclonal
Clone: SPM617
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Non-Human Primates