Order Entry
Poland
ContactUsLinkComponent
Anti-Integrin beta 3 Mouse Monoclonal Antibody [clone: Y2/51]
Anti-Integrin beta 3 Mouse Monoclonal Antibody [clone: Y2/51]
Catalog # ANTIA249079-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-Integrin beta 3 Mouse Monoclonal Antibody [clone: Y2/51]
Catalog # ANTIA249079-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    BDPLT16
  • Antigen symbol:
    Integrin beta 3
  • Clonality:
    Monoclonal
  • Clone:
    Y2/51
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P05106
  • Antigen synonyms:
    HPA 1|CD61 antigen|GP3A|BDPLT2|CD 61|HPA 4|GPIIIa|GT|CD61
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    105 kDa/90 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Phytohemagglutinin (PHA)-stimulated peripheral blood cells
  • Tested applications:
    IHC
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [Y2/51] antibody to Integrin beta 3 for flow cytometry, IF and IHC with samples derived from human.

  • Validated applications: Flow cytometry, IF, IHC
  • Recommended dilutions: Flow cytometry: 0,5 to 1 µg/million cells, IF: 1 to 2 µg/ml, IHC: 1 to 2 µg/ml

Type: Primary
Antigen: Integrin beta 3
Clonality: Monoclonal
Clone: Y2/51
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human