Order Entry
Poland
ContactUsLinkComponent
Anti-Integrin alpha V Mouse Monoclonal Antibody [clone: ITGAV/1610]
Anti-Integrin alpha V Mouse Monoclonal Antibody [clone: ITGAV/1610]
Catalog # ANTIA249070-100
Supplier:  Antibodies.com
New Product
undefined
Anti-Integrin alpha V Mouse Monoclonal Antibody [clone: ITGAV/1610]
Catalog # ANTIA249070-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    antigen identified by monoclonal
  • Antigen symbol:
    Integrin alpha V
  • Clonality:
    Monoclonal
  • Clone:
    ITGAV/1610
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P06756
  • Antigen synonyms:
    Integrin alpha-5|CD 51|DKFZp686A08142|Integrin alpha-V light chain|CD51|integrin alpha V beta 3|Integrin alpha V|integrin alpha-V|Integrin alpha five
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    150 - 160 kDa under non - reducing conditions, Under reducing conditions, cleaved into 2 bands of about 125 - 130 kDa/20 - 25 kDa,
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human Integrin alpha V beta 1 protein
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [ITGAV/1610] antibody to Integrin alpha V for ELISA, flow cytometry, IF and WB with samples derived from human.

  • Validated applications: ELISA, Flow cytometry, IF, WB
  • Recommended dilutions: ELISA: 1 to 5 µg/ml, Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml

Type: Primary
Antigen: Integrin alpha V
Clonality: Monoclonal
Clone: ITGAV/1610
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human