Anti-Interferon gamma Mouse Monoclonal Antibody [clone: G-30]
Catalog # ANTIA248923-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:IFG
- Antigen symbol:Interferon gamma
- Clonality:Monoclonal
- Clone:G-30
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P01579
- Antigen synonyms:IFN immune|IFI|IFN, immune|immune interferon|IFN gamma|IFNG|IFN-gamma|Interferon gamma|IFNG_HUMAN
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:20 - 25 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant human IFN ga mMa
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [G-30] antibody to Interferon gamma for flow cytometry and IF with samples derived from human.
- Validated applications: Flow cytometry, IF
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml
Type: Primary
Antigen: Interferon gamma
Clonality: Monoclonal
Clone: G-30
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human