Order Entry
Poland
ContactUsLinkComponent
Anti-Interferon alpha 1 Mouse Monoclonal Antibody [clone: 2-48]
Anti-Interferon alpha 1 Mouse Monoclonal Antibody [clone: 2-48]
Catalog # ANTIA248917-100
Supplier:  Antibodies.com
New Product
undefined
Anti-Interferon alpha 1 Mouse Monoclonal Antibody [clone: 2-48]
Catalog # ANTIA248917-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    IFL
  • Antigen symbol:
    Interferon alpha 1
  • Clonality:
    Monoclonal
  • Clone:
    2-48
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P01562
  • Antigen synonyms:
    IFNA1|IFNA1_HUMAN|IFN alpha 1b|IFNA13|IFN alpha|IFN|IFN alphaD|IFN-alpha-1/13|IFN alpha 1/13
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    16 - 27 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant human IFN alpha 1
  • Tested applications:
    IHC
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [2-48] antibody to Interferon alpha 1 for flow cytometry, WB, IHC and IF with samples derived from human.

  • Validated applications: Flow cytometry, WB, IHC, IF
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, WB: 1 to 2 µg/ml, IF: 1 to 2 µg/ml

Type: Primary
Antigen: Interferon alpha 1
Clonality: Monoclonal
Clone: 2-48
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human