Order Entry
Poland
ContactUsLinkComponent
 
Anti-Desmocollin 2 Mouse Recombinant Antibody [clone: rDSC2/3437]
  ANTIA248355-100
New Product
undefined
Anti-Desmocollin 2 Mouse Recombinant Antibody [clone: rDSC2/3437]
  ANTIA248355-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    ARVD11
  • Antigen symbol:
    Desmocollin 2
  • Clonality:
    Recombinant
  • Clone:
    rDSC2/3437
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q02487
  • Antigen synonyms:
    CDHF2|Desmosomal glycoprotein II|Desmosomal glycoprotein II and III|Desmocollin 2|Desmocollin-3|Desmocollin-2|Desmocollin 3|Cadherin family member 2|Desmocollin2
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    110 kDa (DSC2)/100 kDa (DSC3)
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human DSC2 protein
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Recombinant mouse monoclonal [rDSC2/3437] antibody to Desmocollin 2 for ELISA with samples derived from human, mouse and rat.

  • Validated applications: ELISA

Type: Primary
Antigen: Desmocollin 2
Clonality: Recombinant
Clone: rDSC2/3437
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat