Anti-Peroxiredoxin 4 Mouse Monoclonal Antibody [clone: CPTC-PRDX4-2]
Catalog # ANTIA248147-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Antioxidant enzyme 372
- Antigen symbol:Peroxiredoxin 4
- Clonality:Monoclonal
- Clone:CPTC-PRDX4-2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q13162
- Antigen synonyms:Peroxiredoxin-4|Peroxiredoxin4|PRDX 4|Antioxidant enzyme AOE372|Peroxiredoxin IV|Peroxiredoxin 4|EC 1.11.1.15|AOE37 2|AOE37-2
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:27 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human PRDX4 protein
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [CPTC-PRDX4-2] antibody to Peroxiredoxin 4 for ELISA with samples derived from human.
- Validated applications: ELISA
Type: Primary
Antigen: Peroxiredoxin 4
Clonality: Monoclonal
Clone: CPTC-PRDX4-2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human