Anti-Kir6.1 Mouse Monoclonal Antibody [clone: S366-60]
ANTIA305001-100
New Product
- Antibody type:Primary
- Antigen symbol:Kir6.1
- Clonality:Monoclonal
- Clone:S366-60
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q63664
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 50% glycerol and 0,1% sodium azid
- Molecular weight:~45 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Fusion protein amino acids 306 - 424 (Cytoplasmic C - terminus) of rat Kir6.1.
- Tested applications:IHC, ICC/IF
- Purification:Protein G purification
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [S366-60] antibody to Kir6.1 for WB, IHC and ICC/IF with samples derived from human, mouse and rat.
- Validated applications: WB, IHC, ICC/IF
- Recommended dilutions: WB: 1:1000, ICC/IF: 1:100
Type: Primary
Antigen: Kir6.1
Clonality: Monoclonal
Clone: S366-60
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human;Mouse;Rat