Anti-Myc Tag Rabbit Monoclonal Antibody [Clone: ARC5004-12]
Catalog # ANTIA305783-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:avian myelocytomatosis viral oncogene homolog
- Antigen symbol:Myc Tag
- Clonality:Monoclonal
- Clone:ARC5004-12
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Species independent
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:c-myc tag|oncogene c-Myc|MRTL|myc proto-oncogene protein|bHLHe39|class E basic helix-loop-helix protein 39|myc-related translation/localization regulatory factor|c-Myc|Myc Epitope Tag
- Storage buffer:Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,05% proclin 300
- Molecular weight:58 (NLK)/72 (YAP1)/55 kDa (PTEN)
- Sequence:EQKLISEEDL
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:A synthetic peptide corresponding to Myc tag.
- Tested applications:ICC/IF
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit monoclonal [ARC5004-12] antibody to Myc Tag for WB, ICC/IF and IP with samples derived from species independent.
- Validated applications: WB, ICC/IF, IP
- Recommended dilutions: WB: 1:10000 to 1:140000, ICC/IF: 1:50 to 1:200, IP: 1:500 to 1:1000
Type: Primary
Antigen: Myc Tag
Clonality: Monoclonal
Clone: ARC5004-12
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Species Independent