- Pk:96 Tests
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:Complement C6
- Description:Human complement C6 ELISA kit
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Cross reactivity:Human Complement C6 ELISA Kit exhibits high specificity and excellent specificity for the detection of human Complement C6. No significant cross-reactivity or interference between Complement C6 and analogues was observed.
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf life:Store for 6 months at 4 °C
- Detection range:0,625 - 40 ng/ml
- Storage temperature:4 °C
- Sample volume:100 μl
- Sensitivity:0,375 ng/ml
- Regulatory status:RUO
Human Complement C6 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Complement C6 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready to use ELISA kit
- Detection range: 0,625 to 40 ng/ml
- Sensitivity: 0,375 ng/ml
- Sample type: Serum, plasma, tissue homogenates and other biological fluids
- Assay precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Human Complement C6 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Complement C6 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Complement C6 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Complement C6 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Complement C6 Antibody, which binds the captured Complement C6 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Complement C6 captured in each well. The concentration of Complement C6 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.