Order Entry
Poland
ContactUsLinkComponent
 
Human Triosephosphate Isomerase ELISA Kit
  ANTIA312498-96
New Product
 :  
undefined
Human Triosephosphate Isomerase ELISA Kit
  ANTIA312498-96
New Product
 :  

 

  • Pk:
    96 Tests
  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    Triosephosphate Isomerase
  • Description:
    Human Triosephosphate Isomerase ELISA kit
  • Sample type:
    Serum, plasma or other biological fluids
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    125 - 2000 ng/L
  • Storage temperature:
    4 °C
  • Sample volume:
    40 μl
  • Sensitivity:
    9,15 ng/L
  • Regulatory status:
    RUO

 

 

Human Triosephosphate Isomerase ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Triosephosphate Isomerase in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 125 to 2000 ng/L
  • Sensitivity: 9,15 ng/L
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Human Triosephosphate Isomerase ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Triosephosphate Isomerase in An antibody specific for Triosephosphate Isomerase has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Triosephosphate Isomerase present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-Triosephosphate Isomerase antibody, which also binds the Triosephosphate Isomerase present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-Triosephosphate Isomerase antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-Triosephosphate Isomerase antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of Triosephosphate Isomerase captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Triosephosphate Isomerase in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.