Specifications
- Pk:96 Tests
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Guinea pig
- Target protein:IgG1
- Description:Guinea pig IgG1 ELISA kit
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Cross reactivity:Guinea pig IgG1 ELISA Kit exhibits high specificity and excellent specificity for the detection of guinea pig IgG1. No significant cross-reactivity or interference between IgG1 and analogues was observed.
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf life:Store for 6 months at 4 °C
- Detection range:1,563 - 100 ng/ml
- Storage temperature:4 °C
- Sample volume:100 μl
- Sensitivity:0,938 ng/ml
- Regulatory status:RUO
Specifications
About this item
Guinea pig IgG1 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of guinea pig IgG1 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Detection Range: 1,563 to 100 ng/ml
- Sensitivity: 0,938 ng/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Guinea Pig IgG1 ELISA Kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of guinea pig IgG1 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for IgG1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the IgG1 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-IgG1 Antibody, which binds the captured IgG1 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of IgG1 captured in each well. The concentration of IgG1 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.