Order Entry
Poland
ContactUsLinkComponent
Mouse Apelin 12 ELISA Kit
Mouse Apelin 12 ELISA Kit
Catalog # ANTIA79007-96
Supplier:  Antibodies.com
New Product
CAS Number:  
undefined
Mouse Apelin 12 ELISA Kit
Catalog # ANTIA79007-96
Supplier:  Antibodies.com
New Product
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    Multiple steps
  • Assay Type:
    Competitive
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    Apelin 12
  • Description:
    Mouse Apelin 12 ELISA kit
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross reactivity:
    Mouse Apelin 12 ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse Apelin 12. No significant cross-reactivity or interference between Apelin 12 and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    1,563 - 100 pg/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    50 μl
  • Sensitivity:
    0,938 pg/ml
  • Regulatory status:
    RUO

Specifications

About this item

Mouse Apelin 12 ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of mouse Apelin 12 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA Kit
  • Detection Range: 1,563 to 100 pg/ml
  • Sensitivity: 0,938 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Mouse Apelin 12 ELISA Kit (A79007) employs the competitive enzyme immunoassay technique for the quantitative measurement of mouse Apelin 12 in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Apelin 12 antigen. During the incubation, Apelin 12 present in the samples or standards competes with the fixed amount of immobilized Apelin 12 for binding sites on the Biotinylated Anti-Apelin 12 Antibody. The more Apelin 12 present in a sample or standard, the less Biotinylated Anti-Apelin 12 Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Apelin 12 Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Apelin 12 present in each sample or standard. The concentration of Apelin 12 can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.