Order Entry
Poland
ContactUsLinkComponent
Mouse VLDL Receptor/VLDL-R ELISA Kit
Mouse VLDL Receptor/VLDL-R ELISA Kit
Catalog # ANTIA310089-96
Supplier:  Antibodies.com
New Product
CAS Number:  
undefined
Mouse VLDL Receptor/VLDL-R ELISA Kit
Catalog # ANTIA310089-96
Supplier:  Antibodies.com
New Product
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    VLDL Receptor/VLDL-R
  • Description:
    Mouse VLDL receptor/VLDL-R ELISA kit
  • Sample type:
    Serum, plasma or other biological fluids
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    1,5 - 24 ng/mL
  • Storage temperature:
    4 °C
  • Sensitivity:
    0,056 ng/ml
  • Regulatory status:
    RUO

Specifications

About this item

Mouse VLDL Receptor/VLDL-R ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse VLDL Receptor/VLDL-R in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 1,5 to 24 ng/mL
  • Sensitivity: 0,056 ng/ml
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Mouse VLDL Receptor/VLDL-R ELISA kit (A310089) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse VLDL Receptor/VLDL-R in An antibody specific for VLDL Receptor/VLDL-R has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the VLDL Receptor/VLDL-R present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-VLDL Receptor/VLDL-R antibody, which also binds the VLDL Receptor/VLDL-R present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-VLDL Receptor/VLDL-R antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-VLDL Receptor/VLDL-R antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. colour develops in proportion to the amount of VLDL Receptor/VLDL-R captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of VLDL Receptor/VLDL-R in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.