Order Entry
Poland
Orders LinkContactUsLinkComponent
Mouse CERK ELISA Kit

Mouse CERK ELISA Kit

Mouse CERK ELISA kit, 96 Tests
Catalog # ANTIA303572-96
Supplier:   Antibodies.com
Mouse CERK ELISA Kit
Catalog # ANTIA303572-96
Supplier:  Antibodies.com
CAS Number:  
Mouse CERK ELISA Kit
Mouse CERK ELISA Kit

Mouse CERK ELISA kit, 96 Tests

CAS Number:  
Price
2 790 zł
Log in for availability
1 * 96 Tests
Product Details & Documents

Mouse CERK ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse CERK in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection Range: 0,156 to 10 ng/ml
  • Sensitivity: 0,094 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra to Assay: CV <8%, Inter to Assay: CV <10%

Mouse CERK ELISA kit (A303572) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse CERK in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for CERK has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the CERK present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated anti-CERK antibody, which binds the captured CERK present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of CERK captured in each well. The concentration of CERK can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Specifications

Specifications

  • Pk:
    96 Tests
  • Description:
    Mouse CERK ELISA kit
  • Primary antibody reactivity:
    Mouse
  • Sample type:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Detection range:
    0,156 - 10 ng/ml
  • Storage temperature:
    4 °C
  • Shelf life:
    Store for 6 months at 4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    4 h 30 min
  • Cross reactivity:
    Mouse CERK ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse CERK. No significant cross-reactivity or interference between CERK and analogues was observed.
  • Assay Principle:
    Quantitative
  • Cat. No.:
    ANTIA303572-96
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    0,094 ng/ml
  • Format:
    Pre-coated
  • Target protein:
    CERK
  • Regulatory status:
    RUO

Specifications