Order Entry
Poland
ContactUsLinkComponent
Pimecrolimus System Suitability Mixture, Reference Standard
Pimecrolimus System Suitability Mixture, Reference Standard
Catalog # 1539111.
Supplier:  USP
New Product
CAS Number:  
Pimecrolimus System Suitability Mixture, Reference Standard
Catalog # 1539111.
Supplier:  USP
New Product
CAS Number:  

Specifications

  • Pk:
    15 mg
  • Standards:
    USP reference standard
  • Description:
    Pimecrolimus system suitability mixture
  • GHS signal:
    Danger

Specifications

About this item