Order Entry
Poland
ContactUsLinkComponent
 
Recombinant DegP from E. coli
New Product
 
Recombinant DegP from E. coli
New Product

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Storage conditions:
    May be stored at 4 °C for short-term only. Aliquot to avoid repeated freezing and thawing. Store at –20 °C. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap. Further dilutions can be made in assay buffer.
  • Gene ID:
    947139
  • Amino acid number:
    27 - 474
  • Protein synonyms:
    htrA|Protease Do|Periplasmic serine endoprotease DegP|Heat shock protein DegP|b0161
  • UniProtKB:
    P0C0V0
  • Protein/peptide name:
    DegP
  • Purity:
    ≥90% (SDS-PAGE)
  • Molecular weight:
    48,8 kD
  • Sequence:
    AETSSATTAQQMPSLAPMLEKVMPSVVSINVEGSTTVNTPRMPRNFQQFFGDDSPFCQEGSPFQSSPFCQGGQGG
    NGGGQQQKFMALGSGVIIDADKGYVVTNNHVVDNATVIKVQLSDGRKFDAKMVGKDPRSDIALIQIQNPKNLTAIK
    MADSDALRVGDYTVAIGNPFGLGETVTSGIVSALGRSGLNAENYENFIQTDAAINRGNSGGALVNLNGELIGINTAIL
    APDGGNIGIGFAIPSNMVKNLTSQMVEYGQVKRGELGIMGTELNSELAKAMKVDAQRGAFVSQVLPNSSAAKAGIK
    AGDVITSLNGKPISSFAALRAQVGTMPVGSKLTLGLLRDGKQVNVNLELQQSSQNQVDSSSIFNGIEGAEMSNKGK
    DQGVVVNNVKTGTPAAQIGLKKGDVIIGANQQAVKNIAELRKVLDSKPSVLALNIQRGDSTIYLLMQ
  • Formulation:
    Supplied as a liquid in 20 mM Tris-HCl, pH 8,0, 50% glycerol
  • Shipping temperature:
    4 °C

 

Frequently Bought Together