Iodine trichloride (Wijs'reagent) for determination of iodine value in fats
Catalog # CHMP116950872.1000
Supplier: CHEMPUR
New Product
CAS Number:
Specifications
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:233.26 g/mol
CAS Number:865-44-1
Formula:ICl₃
MDL Number:MFCD00036291