Optipette Single Channel Pipettes, Starter 4-Pack
Catalog # HTLA7902
Supplier: HTL
Specifications
- Description:Variable volume pipettes Starter 4-pack
- Pk:1
Specifications
About this item
Ergonomically designed Optipette single-channel pipettors feature slim handle, high accuracy and precision rates combined with their tremendous robustness.
- Contoured shape of the handle, equipped with a finger-hook guarantee correct grip reducing wrist strain and fatigue
- Low pipettor weight and soft springs system provides comfort at work, even during long pipetting series
- Reliable sealing system ensures reliability of results
- Built-in user-recalibration system allows matching of the pipette for your specific application
New pipetting mechanism allows for precise and effortless setting of pipettor volume. Winding the counter from minimum to maximum volume can be performed with one hand in minimal time.
Delivery: Package includes OP10, OP20, OP200 and OP1000 pipettes, Plexi 4-position stand, tips 10 μl (96 pieces), tips 200 μl (96 pieces), tips 1000 μl (96 pieces), Calibration tool ×4 and instruction manuals.