Order Entry
Poland
ContactUsLinkComponent
Optipette Single Channel Pipettes, Variable Volume
Optipette Single Channel Pipettes, Variable Volume
Catalog # HTLA5602
Supplier:  HTL
Optipette Single Channel Pipettes, Variable Volume
Catalog # HTLA5602
Supplier:  HTL

Specifications

  • Colour code:
    Red
  • Volume:
    0,5 - 10 µl
  • Description:
    OP10 Variable volume pipette fits 10 µl standard tips
  • Seq:
    01
  • Accuracy:
    ±4 - ±0,5 %
  • Imprecision:
    ±2,8 - ±0,4 %
  • Pk:
    1

Specifications

About this item

Ergonomically designed Optipette single-channel pipettors feature slim handle, high accuracy and precision rates combined with their tremendous robustness.

  • Contoured shape of the handle, equipped with a finger-hook guarantee correct grip reducing wrist strain and fatigue
  • Low pipettor weight and soft springs system provides comfort at work, even during long pipetting series
  • Reliable sealing system ensures reliability of results
  • Built-in user-recalibration system allows matching of the pipette for your specific application

New pipetting mechanism allows for precise and effortless setting of pipettor volume. Winding the counter from minimum to maximum volume can be performed with one hand in minimal time.