Order Entry
Poland
ContactUsLinkComponent
Crystal violet, Sigma-Aldrich®
Crystal violet, Sigma-Aldrich®
Catalog # SIAL61135-25G
Supplier:  Merck
CAS Number:  
Crystal violet, Sigma-Aldrich®
Catalog # SIAL61135-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :407.99 g/mol
Merck Index:13,04406
Melting Pt:189…194 °C
Density:1.19 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-953-6
UN:3077
ADR:9,III
 :M-Clarity™ quality level = MQ200
 :GHS05
 :GHS08
 :GHS07
 :GHS09
CAS Number:548-62-9
Formula:C₂₅H₃₀ClN₃
MDL Number:MFCD00011750