Crystal violet 90+% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:548-62-9
:
Formula:C₂₅H₃₀ClN₃
MDL Number:MFCD00011750
:407.99 g/mol
Merck Index:14,4395
Melting Pt:∼ 215 °C (dec.)
Density:1.19 g/cm³ (20 °C)
EINECS:208-953-6
UN:3077
ADR:9,III
:GHS05
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Related Information
Frequently Bought Together