Order Entry
Poland
ContactUsLinkComponent
VWR®, Organic reference standard, Aroclor 1242 (PCB-mixture, 42% chlorinated) 1000 µg/ml in hexane
VWR®, Organic reference standard, Aroclor 1242 (PCB-mixture, 42% chlorinated) 1000 µg/ml in hexane
Catalog # 122792T
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Organic reference standard, Aroclor 1242 (PCB-mixture, 42% chlorinated) 1000 µg/ml in hexane
Catalog # 122792T
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass ampoule
  • Pk:
    1 mL
  • Description:
    Aroclor 1242 (PCB-mixture, 42% chlorinated) 1,000 µg/ml in hexane

Specifications

About this item