Order Entry
Poland
Orders LinkContactUsLinkComponent
Fuchsin (acid), pure, certified

Fuchsin (acid), pure, certified

Fuchsin (acid), pure, certified
Certificates
Fuchsin (acid), pure, certified

2 Options Available Below

Product Details & Documents
Synonyms:  Rubin S, CN DcR-8, Acid Violet 19, Acid Fuchsin, Acid Fuchsin sodium salt, Fuchsin S, CI 42685, Acid Magenta, 3-(1-(4-Amino-3-methyl-5-sulphonatophenyl)-1-(4-amino-3-sulphonatophenyl)methylene)cyclohexa-1,4-dienesulphonic acid, Benzenesulphonic acid, 2-amino-5-[(4-amino-3-sulphophenyl)(4-imino-3-sulpho-2,5-cyclohexadien-1-ylidene)methyl]-3-methyl-, sodium salt (1:2)
  • CAS Number:3244-88-0
  • Formula:C₂₀H₁₇N₃Na₂O₉S₃
  • MDL Number:MFCD00013286
  • Molecular Weight:585.55 g/mol
  • Merck Index:12,00109
  • Melting Pt:? 130 °C
  • Storage Temperature:Ambient
  • EINECS:221-816-5

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Dark green Crystalline powder
  • Infrared spectrum:
    Conforms
  • Dye content:
    ≥60 %
  • UV Lambda max:
    540 to 549 nm (10 mg/l in water + 2.5ml 0.1N HCl)
  • Absorption Ratio P-15/P+15:
    1.10 to 1.26
  • BSC Certification:
    Passes
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.