Order Entry
Poland
ContactUsLinkComponent
Thiamine hydrochloride, Sigma-Aldrich®
Thiamine hydrochloride, Sigma-Aldrich®
Catalog # SIAL04573-50G-F
Supplier:  Merck
CAS Number:  
Thiamine hydrochloride, Sigma-Aldrich®
Catalog # SIAL04573-50G-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :337.27 g/mol
Merck Index:13,09366
Melting Pt:252 °C
Density:1.401 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:200-641-8
CAS Number:67-03-8
Formula:C₁₂H₁₈Cl₂N₄OS
MDL Number:MFCD00012780