
Aluminium trilactate 90-95% contains ≈5% water
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:18917-91-4
:MFCD00043086
Formula:(H₃CCH(OH)COO)₃Al
MDL Number:MFCD20529175
:294.19 g/mol
Merck Index:14,348
Melting Pt:> 300 °C
EINECS:242-670-9
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together