Order Entry
Poland
ContactUsLinkComponent
Anti-PRLR Mouse Monoclonal Antibody [clone: ERSV8-1]
Anti-PRLR Mouse Monoclonal Antibody [clone: ERSV8-1]
Catalog # PRSI33-926
Supplier:  ProSci Inc.
Anti-PRLR Mouse Monoclonal Antibody [clone: ERSV8-1]
Catalog # PRSI33-926
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    prolactin receptor
  • Antigen symbol:
    PRLR
  • Clonality:
    Monoclonal
  • Clone:
    ERSV8-1
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Format:
    Purified Antibody
  • Form:
    Liquid
  • Gene ID:
    P16471
  • Antigen synonyms:
    PRLR
  • Storage buffer:
    PBS with 0,1 mg/ml BSA and 0,05% sodium azide
  • Storage temperature:
    Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
  • Concentration:
    0,2 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    Recombinant human Prolactin receptor was used as the immunogen for this antibody.
  • Purification:
    Protein G affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Prolactin is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. The initial step in its action is the binding to a specific membrane receptor, which belongs to the superfamily of class 1 cytokine receptors. The function of the prolactin receptor is mediated, at least in part, by two families of signaling molecules: Janus kinases and signal transducers and activators of transcription.

IF: 0,5 - 1 µg/ml IHC (FFPE): 0,5 - 1 µg/ml for 30 minutes at RT (1) Prediluted format: incubate for 30 min at RT (2) The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the Prolactin receptor antibody to be titered up or down for optimal performance.

1. Staining of formalin/paraffin tissues requires digestion of tissue sections with pepsin at 1mg/ml Tris-HCl, pH 2.0 for 15 min at RT or 10 min at 37oC.
2. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.

Type: Primary
Antigen: PRLR
Clonality: Monoclonal
Clone: ERSV8-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human