Order Entry
Poland
ContactUsLinkComponent
Anti-S100A1 Mouse Monoclonal Antibody [clone: S1-61]
Anti-S100A1 Mouse Monoclonal Antibody [clone: S1-61]
Catalog # PRSI33-750
Supplier:  ProSci Inc.
Anti-S100A1 Mouse Monoclonal Antibody [clone: S1-61]
Catalog # PRSI33-750
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    S100 calcium binding protein A1
  • Antigen symbol:
    S100A1
  • Clonality:
    Monoclonal
  • Clone:
    S1-61
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Format:
    Purified Antibody
  • Form:
    Liquid
  • Gene ID:
    P23297
  • Antigen synonyms:
    S100A1
  • Storage buffer:
    PBS with 0,1 mg/ml BSA and 0,05% sodium azide
  • Storage temperature:
    Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
  • Concentration:
    0,2 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    Purified human S100A protein was used as the immunogen for this S100 antibody.
  • Purification:
    Protein G affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

S100 belongs to the family of calcium binding proteins. S100A and S100B proteins are two members of the S100 family. S100A is composed of an alpha and beta chain whereas S100B is composed of two beta chains. S100 protein has been found in normal melanocytes, Langerhans cells, histiocytes, chondrocytes, lipocytes, skeletal and cardiac muscle, Schwann cells, epithelial and myoepithelial cells of the breast, salivary and sweat glands, as well as in glial cells. Neoplasms derived from these cells also express S100 protein, albeit non-uniformly. A large number of well-differentiated tumors of the salivary gland, adipose and cartilaginous tissue, and Schwann cell-derived tumors express S100 protein. Almost all malignant melanomas and cases of histiocytosis X are positive for S100 protein. Despite the fact that S100 protein is an ubiquitous substance, its demonstration is of great value in the identification of several neoplasms, particularly melanomas and their metastases.

Immunofluorescence: 1 - 2 µg/ml Immunohistochemistry (FFPE): 1 - 2 µg/ml for 30 min at RT The optimal dilution of the S100A antibody for each application should be determined by the researcher.

Type: Primary
Antigen: S100A1
Clonality: Monoclonal
Clone: S1-61
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human