Specifications
- Antibody type:Primary
- Antigen name:complement component 5a receptor 2
- Antigen symbol:C5ar2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Primates,Human
- Western blot:Yes
- Format:Purified Antibody
- Form:Liquid
- Gene ID:Q695P6
- Antigen synonyms:C5ar2
- Storage buffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Storage temperature:Aliquot and Store at –20 °C. Avoid freez-thaw cycles.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:A recombinant protein fragment from the N-terminal region of human GPR77 was used as the immunogen for this antibody.
- Purification:Protein A Affinity Chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
G protein receptors, also called GPRs, GPCRs, and seven-transmembrane domain receptors, are a large family of receptors with possibly over 800 members. These receptors sense stimuli outside, convey the signal across the cell membrane, and activate internal signal transduction pathways and cellular responses. GPR77 is involved in the complement system of the innate immune response and may signal through the beta-arrestin pathway, and possibly act as a decoy receptor. It is co-expressed with the C5a receptor on neutrophils and may modulate C5a1 activity.
Western blot: 1-3 µg/ml Provided assay concentrations are sµggestions only, GPR77 antibody titration may be required for optimal results.
Type: Primary
Antigen: C5ar2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Primate