Anti-SERPINA6 Rabbit Polyclonal Antibody
numer katalogowy PRSI15-102
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Corticosteroid-binding globulin
- Antigen symbol:SERPINA6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:866
- Antigen synonyms:SERPINA6|Corticosteroid-binding globulin|Serpin A6|Transcortin|CBG
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 136-405 of human SERPINA6 (NP_001747.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SERPINA6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat