Anti-FIBP Rabbit Polyclonal Antibody
Catalog # PRSI15-076
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Acidic fibroblast growth factor intracellular-binding protein
- Antigen symbol:FIBP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:9158
- Antigen synonyms:FIBP|Acidic fibroblast growth factor intracellular-binding protein|aFGF intracellular-binding protein|FGF-1 intracellular-binding protein
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-364 of human FIBP (NP_004205.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: FIBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse