Anti-USP7 Rabbit Polyclonal Antibody
Catalog # PRSI15-072
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Ubiquitin specific peptidase 7 (herpes virus-associated)
- Antigen symbol:USP7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:7874
- Antigen synonyms:TEF1|HAUSP|Deubiquitinating enzyme 7|Ubiquitin thioesterase 7|USP7|Ubiquitin specific protease 7
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 206-1102 of human HAUSP/USP7 (NP_003461.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: USP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat