Anti-WNT2 Rabbit Polyclonal Antibody
Catalog # PRSI15-071
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Wnt family member 2
- Antigen symbol:WNT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:7472
- Antigen synonyms:epididymis secretory sperm binding protein|Int-1-related protein|INT1L1|int-1-like protein 1|IRP|wingless-type MMTV integration site family member 2|secreted growth factor|protein Wnt-2
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 1-100 of human WNT2 (NP_003382.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: WNT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat